5-(2-Pyridyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-(1H-Tetrazol-5-yl)pyridine, 5-(2-Pyridyl)-1H-tetrazole
T3431-1G
T3431-5G
TCT3431-1GEA
119.48
USD
TCT3431-1G
TCT3431-5G
5-(2-Pyridyl)-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-(2-Pyridyl)-1H-tetrazole
Formula:
C₆H₅N₅ MW: 147.14 g/mol |
MDL Number:
MFCD00068114 CAS Number: 33893-89-9 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...