4-(Trifluoromethyl)benzoic acid hydrazide ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-(Trifluoromethyl)benzoylhydrazine, 4-(Trifluoromethyl)benzhydrazide, 4-(Trifluoromethyl)benzoic Hydrazide, 4-(Trifluoromethyl)benzohydrazide

T3372-1G T3372-5G
TCT3372-1GEA 63.86 USD
TCT3372-1G TCT3372-5G
4-(Trifluoromethyl)benzoic acid hydrazide ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)benzoic acid hydrazide
Formula: C₈H₇F₃N₂O
MW: 204.15 g/mol
MDL Number: MFCD00051703
CAS Number: 339-59-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d336c8ab321cb610277777c3e4b33c337cde768&vid=c8ed6eb4dd219e7522c1eb2dfdbfeed2e3c9f082&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Bromination Method) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR