Tris(3-methoxyphenyl)phosphine ≥98.0% (by HPLC)

Supplier: TCI America
Warning

T2946-25G T2946-5G
TCT2946-25GEA 212.18 USD
TCT2946-25G TCT2946-5G
Tris(3-methoxyphenyl)phosphine ≥98.0% (by HPLC)
Tris(3-methoxyphenyl)phosphine
Formula: C₂₁H₂₁O₃P
MDL Number: MFCD00008386
CAS Number: 29949-84-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27bd9ea5ae7d332b9f56c875dd35ca59b9f8b5d9&vid=d20392ed0562662e9aad64fdf92f0fd151bab512&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Iodometric Titration) min. 95.0 %
Melting point 113.0 to 117.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR