Dihydroxybis(hydrogen Lactato)titanium(IV) ∼44% in 2-propanol/water

Supplier: TCI America
Danger

Synonyms: Titanium(IV) bis(hydrogen Lactato)dihydroxide

T2612-25G T2612-500G
TCT2612-25GEA 30.9 USD
TCT2612-25G TCT2612-500G
Dihydroxybis(hydrogen Lactato)titanium(IV) ∼44% in 2-propanol/water
Dihydroxybis(hydrogen Lactato)titanium(IV)
Formula: C₆H₁₂O₈Ti
MW: 260.02 g/mol
CAS Number: 79110-90-0
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=059153c0557bae12ae472e68f0803e69cf8b47c1&vid=5fcd4de3600e85d3f3b6ef776181ed438f56fddb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Concentration (Gravimetric Method) 42.0 to 46.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR