Phenylbis(phenylthio)methane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (Phenylmethylene)bis(phenylsulphane), alpha,alpha-Bis(phenylthio)toluene, Benzaldehyde Diphenyl Dithioacetal

P2764-1G P2764-5G
TCP2764-1GEA 43.26 USD
TCP2764-1G TCP2764-5G
Phenylbis(phenylthio)methane ≥98.0% (by GC)
Phenylbis(phenylthio)methane
Formula: C₁₉H₁₆S₂
MW: 308.46 g/mol
Boiling Pt: 184 °C
MDL Number: MFCD00092116
CAS Number: 7695-69-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR