2-(Propylamino)ethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

P2743-25G P2743-5G
TCP2743-25GEA 329.6 USD
TCP2743-25G TCP2743-5G
2-(Propylamino)ethanol ≥98.0% (by GC, titration analysis)
2-(Propylamino)ethanol
Formula: C₅H₁₃NO
Boiling Pt: 182 °C
Flash Pt: 78 °C
MDL Number: MFCD00002844
CAS Number: 16369-21-4
EINECS: 240-426-6
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR