5-Phenyl-1H-[1,2,4]triazol-3-ylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P2658-1G P2658-5G
TCP2658-1GEA 182.31 USD
TCP2658-1G TCP2658-5G
5-Phenyl-1H-[1,2,4]triazol-3-ylamine ≥98.0% (by GC, titration analysis)
5-Phenyl-1H-[1,2,4]triazol-3-ylamine
Formula: C₈H₈N₄
MDL Number: MFCD00086332
CAS Number: 4922-98-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR