Potassium(2-naphthalene)trifluoroborate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

P2655-25G P2655-5G
TCP2655-25GEA 303.85 USD
TCP2655-25G TCP2655-5G
Potassium(2-naphthalene)trifluoroborate ≥98.0% (by HPLC)
Potassium(2-naphthalene)trifluoroborate
Formula: C₁₀H₇BF₃K
MDL Number: MFCD06659941
CAS Number: 668984-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=435af1b8e77dfebe3ca2c9a325f450a05c8196f4&vid=c415b3a04f9ab6b25e1f5cf8fb3ffcb88e693574&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR