Pentachlorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

P2643-5G P2643-25G
TCP2643-5GEA 76.22 USD
TCP2643-5G TCP2643-25G
Pentachlorobenzonitrile ≥98.0% (by GC)
Pentachlorobenzonitrile
Formula: C₇Cl₅N
MDL Number: MFCD00173666
CAS Number: 20925-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64ded6317f8b05bd8e43f22f9b93051a6f428689&vid=8bd42c522a5eefd88215ab0669d57f44bee4c8c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 215.0 to 219.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR