Potassium ethyltrifluoroborate ≥97.0% (W)

Supplier: TCI America
Warning

P2628-1G P2628-5G
TCP2628-1GEA 51.5 USD
TCP2628-1G TCP2628-5G
Potassium ethyltrifluoroborate ≥97.0% (W)
Potassium ethyltrifluoroborate
Formula: C₂H₅BF₃K
MDL Number: MFCD04112713
CAS Number: 44248-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efcf72dca2f27a3ca9189fd8fa59b22beb0881a3&vid=d9d9d0b2b70ca5ed3fcee36df13c7dc30eaf0b82&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Gravimetric) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR