1-Phenyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Phenylpyrazole-4-carboxaldehyde, 1-Phenyl-1H-pyrazole-4-carboxaldehyde

P2473-1G P2473-5G
TCP2473-1GEA 92.7 USD
TCP2473-1G TCP2473-5G
1-Phenyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
1-Phenyl-1H-pyrazole-4-carbaldehyde
Formula: C₁₀H₈N₂O
MW: 172.18 g/mol
MDL Number: MFCD02179568
CAS Number: 54605-72-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR