2-Propylthiophene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-n-Propylthiophene

P2451-25G P2451-5G
TCP2451-25GEA 175.1 USD
TCP2451-25G TCP2451-5G
2-Propylthiophene ≥98.0% (by GC)
2-Propylthiophene
Formula: C₇H₁₀S
MW: 126.22 g/mol
Boiling Pt: 160 °C
Flash Pt: 44 °C
MDL Number: MFCD00014533
CAS Number: 1551-27-5
EINECS: 216-288-8
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2caedc0eccbfe7594fb653e592ad787301ad5fec&vid=14636882ee833eaf1f358c47ad0042549a937605&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR