trans,trans-4'-Pentyl-4-bicyclohexylmethanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: ((trans,trans)-4'-Pentyl-[1,1'-bi(cyclohexan)]-4-yl)methanol, trans,trans-4'-Amyl-4-bicyclohexylmethanol

P2433-1G P2433-5G
TCP2433-1GEA 47.38 USD
TCP2433-1G TCP2433-5G
trans,trans-4'-Pentyl-4-bicyclohexylmethanol ≥98.0% (by GC)
trans,trans-4'-Pentyl-4-bicyclohexylmethanol
Formula: C₁₈H₃₄O
MW: 266.47 g/mol
MDL Number: MFCD16036281
CAS Number: 82598-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=496ed0f8972e0e8bc7159b736437a3a82e842782&vid=b3958cd4776acfddc1f55194fbe666b311fa7d00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR