4-(Prop-2-yn-1-yl)morpholine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-(2-Propyn-1-yl)morpholine, 4-Propargylmorpholine
P2170-200MG
P2170-1G
TCP2170-200MGEA
120.51
USD
TCP2170-200MG
TCP2170-1G
4-(Prop-2-yn-1-yl)morpholine ≥98.0% (by GC)
4-(Prop-2-yn-1-yl)morpholine
Formula:
C₇H₁₁NO MW: 125.17 g/mol Boiling Pt: 186 °C |
MDL Number:
MFCD00510919 CAS Number: 5799-76-8 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...