Sodium Pyrimidine-2-sulfinate ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Sodium pyrimidine-2-sulphinate

O0541-1G O0541-250MG
TCO0541-1GEA 545.9 USD
TCO0541-1G TCO0541-250MG
Sodium Pyrimidine-2-sulfinate ≥97.0% (by HPLC, titration analysis)
Sodium Pyrimidine-2-sulfinate
Formula: C₄H₃N₂NaO₂S
MW: 166.13 g/mol
MDL Number: MFCD32068077
CAS Number: 2188151-68-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR