Sodium 4-phenylbutyrate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Sodium 4-phenylbutanoate, 4-Phenylbutyric acid sodium salt

O0511-1G O0511-5G
TCO0511-1GEA 96.82 USD
TCO0511-1G TCO0511-5G
Sodium 4-phenylbutyrate ≥98.0% (by HPLC, titration analysis)
Sodium 4-phenylbutyrate
Formula: C₁₀H₁₁O₂·Na
MW: 186.2 g/mol
MDL Number: MFCD00800247
CAS Number: 1716-12-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR