Nifuratel ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 5-[(Methylthio)methyl]-3-[[(5-nitro-2-furanyl)methylene]amino]-2-oxazolidinone, 5-((Methylthio)methyl)-3-(((5-nitrofuran-2-yl)methylene)amino)oxazolidin-2-one

N1167-25G N1167-5G
TCN1167-25GEA 251.32 USD
TCN1167-25G TCN1167-5G
Nifuratel ≥98.0% (by HPLC)
Nifuratel
Formula: C₁₀H₁₁N₃O₅S
MW: 285.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00057257
CAS Number: 4936-47-4
EINECS: 225-576-2
Merck Index: 14,06532

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 14.00 to 15.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR