Neobavaisoflavone ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 7-Hydroxy-3-(4-hydroxy-3-(3-methylbut-2-en-1-yl)phenyl)-4H-chromen-4-one, 7-Hydroxy-3-[4-hydroxy-3-(3-methyl-2-buten-1-yl)phenyl]-4H-1-benzopyran-4-one, 4',7-Dihydroxy-3'-(3-methyl-2-butenyl)isoflavone

N1122-25MG N1122-5MG
TCN1122-25MGEA 371.83 USD
TCN1122-25MG TCN1122-5MG
Neobavaisoflavone ≥97.0% (by HPLC)
Neobavaisoflavone
Formula: C₂₀H₁₈O₄
MW: 322.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00800660
CAS Number: 41060-15-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1247b718d94fefa344947993c166a709f99d0f7d&vid=485a92d7341bc0bdfc626be1b0fd865817ab4bc0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Carbon) 72.00 to 76.50 %
Melting point 195.0 to 199.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR