NBD-556 ≥98.0% (by HPLC)

Supplier: TCI America
Danger

Synonyms: N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethylpiperidin-4-yl)oxalamide, N1-(4-Chlorophenyl)-N2-(2,2,6,6-tetramethyl-4-piperidinyl)ethanediamide

N1090-100MG N1090-25MG
TCN1090-100MGEA 739.54 USD
TCN1090-100MG TCN1090-25MG
NBD-556 ≥98.0% (by HPLC)
NBD-556
Formula: C₁₇H₂₄ClN₃O₂
MW: 337.85 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00469538
CAS Number: 333353-44-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=29764c2156a8b8bfef3025db131673b1315253a9&vid=061925ea2f9c81b49454e47371dfa183108ee341&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 11.70 to 13.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR