4-Methyl-4-(methylthio)-2-pentanone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-Methyl-4-methylthio-2-pentanone, 4-Methyl-4-(methylthio)pentan-2-one

M3272-100G M3272-25G
TCM3272-100GEA 166.86 USD
TCM3272-100G TCM3272-25G
4-Methyl-4-(methylthio)-2-pentanone ≥98.0% (by GC)
4-Methyl-4-(methylthio)-2-pentanone
Formula: C₇H₁₄OS
MW: 146.25 g/mol
Boiling Pt: 78 °C
MDL Number: MFCD00049253
CAS Number: 23550-40-5
EINECS: 245-727-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR