1-Methylcyclopentyl acrylate ≥98.0% (by GC) stabilized

Supplier: TCI America


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M3261-1G M3261-5G
TCM3261-1GEA 76.22 USD
TCM3261-1G TCM3261-5G
1-Methylcyclopentyl acrylate ≥98.0% (by GC) stabilized
1-Methylcyclopentyl acrylate
Formula: C₉H₁₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08704215
CAS Number: 178889-49-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR