Methyl-2-(1H-indol-3-yl)-2-oxoacetate ≥96.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 3-Indoleglyoxylate, Methyl indole-3-glyoxylate, Methyl 2-(1H-indol-3-yl)-2-oxoacetate, 2-(1H-Indol-3-yl)-2-oxoacetic acid methyll ester, 3-Indoleglyoxylic acid methyll ester, Methyl 2-(1H-Indol-3-yl)-2-oxoacetate

M3254-1G M3254-5G
TCM3254-1GEA 49.44 USD
TCM3254-1G TCM3254-5G
Methyl-2-(1H-indol-3-yl)-2-oxoacetate ≥96.0% (by GC)
Methyl-2-(1H-indol-3-yl)-2-oxoacetate
Formula: C₁₁H₉NO₃
MW: 203.1941 g/mol
MDL Number: MFCD00047173
CAS Number: 18372-22-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 228.0 to 232.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR