4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-[(4-MethoxyCarbonylmethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid methyll ester, Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate, Methyl 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetate, 4-(MethoxyCarbonylmethyl)phenylboronic acid Pinacol ester

M3236-1G M3236-5G
TCM3236-1GEA 71.07 USD
TCM3236-1G TCM3236-5G
4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Methoxycarbonylmethyl)phenylboronic acid pinacol ester
Formula: C₁₅H₂₁BO₄
MW: 276.1358 g/mol
MDL Number: MFCD09997797
CAS Number: 454185-98-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f14278d2b758fb8ef04cf23d653687c684b45b4&vid=985b5c1157fcbe1170a992b52249dd329d2bdfcb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR