1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
2-[1-(2-Methoxyethyl)-1H-pyrazol-4-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid Pinacol ester, 1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester
M3232-1G
M3232-200MG
TCM3232-1GEA
170.98
USD
TCM3232-1G
TCM3232-200MG
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98.0% (by GC, titration analysis)
1-(2-Methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formula:
C₁₂H₂₁BN₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD12405649 CAS Number: 847818-71-7 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...