2-Methoxy-6-methylbenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
M3208-1G
M3208-5G
TCM3208-1GEA
82.4
USD
TCM3208-1G
TCM3208-5G
2-Methoxy-6-methylbenzoic acid ≥98.0% (by GC, titration analysis)
2-Methoxy-6-methylbenzoic acid
Formula:
C₉H₁₀O₃ |
MDL Number:
MFCD00463871 CAS Number: 6161-65-5 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 139.0 to 143.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...