5-Methoxypyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M3181-1G M3181-200MG
TCM3181-1GEA 215.27 USD
TCM3181-1G TCM3181-200MG
5-Methoxypyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Methoxypyrazine-2-carboxylic acid
Formula: C6H6N2O3
MDL Number: MFCD10697773
CAS Number: 40155-42-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6e1a920d54713358243de43f0c16dc78697fe2b&vid=b81b4496abdb801bd913e9b9fe7c540727ddf1c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point (Decomposition) 196.0 to 200.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR