SMER-3 ≥97.0% (by HPLC)

Supplier: TCI America

Synonyms: 9H-Indeno[1,2-b][1,2,5]oxadiazolo[3,4-e]pyrazin-9-one

M3172-10MG M3172-50MG
TCM3172-10MGEA 128.75 USD
TCM3172-10MG TCM3172-50MG
SMER-3 ≥97.0% (by HPLC)
SMER-3
Formula: C₁₁H₄N₄O₂
MW: 224.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00451458
CAS Number: 67200-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c25b2497b431ac3871fa1be33027eb465b72a5f7&vid=758c1bca2f15314741ff43a58bc1cc723b41a292&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 23.70 to 26.00 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR