5-Methoxyoxindole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 5-Methoxy-2-indolinone, 5-Methoxyindolin-2-one

M3157-1G M3157-200MG
TCM3157-1GEA 175.1 USD
TCM3157-1G TCM3157-200MG
5-Methoxyoxindole ≥98.0% (by GC)
5-Methoxyoxindole
Formula: C9H9NO2
MW: 163.1733 g/mol
MDL Number: MFCD01860251
CAS Number: 7699-18-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a93405b973e7ecc420c437f3fdc780ca3a0287dc&vid=8c4f2d1a4d9be487025ab7c6b24830c9e1fb44ad&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR