4-(Methylsulfonyl)-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
M3146-25G
M3146-5G
TCM3146-25GEA
59.74
USD
TCM3146-25G
TCM3146-5G
4-(Methylsulfonyl)-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
4-(Methylsulphonyl)-2-nitrobenzoic acid
Formula:
C₈H₇NO₆S |
MDL Number:
MFCD01941299 CAS Number: 110964-79-9 |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 215.0 to 219.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...