7-Methoxy-2,3-dihydro-1H-inden-1-one ≥98.0% (by GC)

Supplier: TCI America

M3126-1G M3126-200MG
TCM3126-1GEA 199.82 USD
TCM3126-1G TCM3126-200MG
7-Methoxy-2,3-dihydro-1H-inden-1-one ≥98.0% (by GC)
7-Methoxy-2,3-dihydro-1H-inden-1-one
Formula: C₁₀H₁₀O₂
Boiling Pt: 110 °C
MDL Number: MFCD07784339
CAS Number: 34985-41-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c03257689b23698ebc4735bd4a32bbbec32e4b99&vid=a6260a3af009f180973d5f8635676701ab2362d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR