Methyl-2-fluoro-5-nitrobenzenecarboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methyl 2-fluoro-5-nitrobenzenecarboxylate, Methyl 2-fluoro-5-nitrobenzoate

M3115-25G M3115-5G
TCM3115-25GEA 234.84 USD
TCM3115-25G TCM3115-5G
Methyl-2-fluoro-5-nitrobenzenecarboxylate ≥98.0% (by GC)
Methyl-2-fluoro-5-nitrobenzenecarboxylate
Formula: C₈H₆FNO₄
MW: 199.14 g/mol
Boiling Pt: 139 °C
MDL Number: MFCD06203689
CAS Number: 2965-22-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e8128100b4549d3136e4c1f9d3a9ec339c9ccb3&vid=f17e33fd403195402f33681c7669a0a0d2c97aa0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR