4-Methyl-2-(p-tolyl)pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Methyl-2-(4-methylphenyl)pyridine

M3105-1G M3105-200MG
TCM3105-1GEA 219.39 USD
TCM3105-1G TCM3105-200MG
4-Methyl-2-(p-tolyl)pyridine ≥98.0% (by GC, titration analysis)
4-Methyl-2-(p-tolyl)pyridine
Formula: C₁₃H₁₃N
MW: 183.25 g/mol
MDL Number: MFCD09031945
CAS Number: 80635-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78f9a0b32c6efb2d716dc5cb8f281867308a7a0a&vid=0cd42268136900f9a3b22f7aaa9f344cd2516c2c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR