Sophoridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: (7aR,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, (5beta)-Matridin-15-one, (41S,7aR,13aR,13bR)-Dodecahydro-1H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10(41H)-one

M3093-250MG M3093-50MG
TCM3093-250MGEA 225.57 USD
TCM3093-250MG TCM3093-50MG
Sophoridine ≥98.0% (by GC, titration analysis)
Sophoridine
Formula: C₁₅H₂₄N₂O
MW: 248.37 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00238686
CAS Number: 6882-68-4
UN: 1544
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=396736a50ec037fefa2fab78fa83f685d8c7774d&vid=a835cd78383b2b3d3dc90f54c207799f88a1405c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C
Specific rotation [a]20/D -66.0 to -73.0 deg (C=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR