Meisoindigo ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Methylisoindigotin, 3-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)-1,3-dihydro-1-methyl-2H-indol-2-one, 1-Methyl-[3,3'-biindolinylidene]-2,2'-dione

M3082-100MG M3082-25MG
TCM3082-100MGEA 422.3 USD
TCM3082-100MG TCM3082-25MG
Meisoindigo ≥98.0% (by HPLC)
Meisoindigo
Formula: C₁₇H₁₂N₂O₂
MW: 276.29 g/mol
Melting Pt: 230 °C
MDL Number: MFCD05988503
CAS Number: 97207-47-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d2751525137059d15e2ba8fdcdf0bf5898f7dc&vid=81a96545090b384f3cd2155f95ceb147d716d10b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Brown to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.50 to 10.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR