1,1'-[Methylenebis(2,6-diethyl-4,1-phenylene)]bis[3-(2-nitrophenyl)urea] ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

M3072-1G M3072-5G
TCM3072-1GEA 125.66 USD
TCM3072-1G TCM3072-5G
1,1'-[Methylenebis(2,6-diethyl-4,1-phenylene)]bis[3-(2-nitrophenyl)urea] ≥98.0% (by HPLC, total nitrogen)
1,1'-[Methylenebis(2,6-diethyl-4,1-phenylene)]bis[3-(2-nitrophenyl)urea]
Formula: C₃₅H₃₈N₆O₆
CAS Number: 2414546-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98d7bca398e3115a982c5e04987ab293c4bcedc5&vid=9d94713925ce5c93b0be00b88c25bcd38b090a39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR