5-Methylpyrazin-2-amine ≥99.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Amino-5-methylpyrazine

M3006-1G M3006-5G
TCM3006-1GEA 90.64 USD
TCM3006-1G TCM3006-5G
5-Methylpyrazin-2-amine ≥99.0% (by GC)
5-Methylpyrazin-2-amine
Formula: C5H7N3
MW: 109.13 g/mol
MDL Number: MFCD08437664
CAS Number: 5521-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc3a5dad81dd7d38cc240679d87d673f356f032b&vid=2537d68b82da6a5a2a9bae6735822e1b19f48dd1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR