Methyl 2-Nitrophenyl sulfone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-(Methylsulphonyl)-2-nitrobenzene

M2927-1G M2927-5G
TCM2927-1GEA 105.06 USD
TCM2927-1G TCM2927-5G
Methyl 2-Nitrophenyl sulfone ≥98.0% (by GC)
Methyl 2-Nitrophenyl sulfone
Formula: C₇H₇NO₄S
MW: 201.20 g/mol
MDL Number: MFCD12828669
CAS Number: 2976-34-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53dd1e9d9bd15e6213c5737978680291fa7f69c1&vid=922bd273c30a66ad7b2494d5b41528248499872b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR