Methyl-5-bromopyrimidine-2-carboxyate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 5-Bromopyrimidine-2-carboxylate, 5-Bromopyrimidine-2-carboxylic acid methyll ester

M2910-1G M2910-200MG
TCM2910-1GEA 103 USD
TCM2910-1G TCM2910-200MG
Methyl-5-bromopyrimidine-2-carboxyate ≥98.0% (by GC)
Methyl-5-bromopyrimidine-2-carboxyate
Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Melting Pt: 149 °C
MDL Number: MFCD11111606
CAS Number: 89581-38-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17d46893b6665f616764f0846172282849c689d9&vid=1083f5b0eb3ec7e3f31f032671cf08386cc73fe0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR