2-Nitro-5-picoline ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Methyl-2-nitropyridine, 6-Nitro-3-picoline

M2890-1G M2890-5G
TCM2890-1GEA 46.35 USD
TCM2890-1G TCM2890-5G
2-Nitro-5-picoline ≥98.0% (by GC)
2-Nitro-5-picoline
Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
MDL Number: MFCD00234182
CAS Number: 1074-38-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f82faf33f660b7097466853e7d9deda6f16b539&vid=d36f05897454938149d059144c6fa8b5ecdfa929&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR