4-(Methylthio)phenylacetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2874-1G M2874-5G
TCM2874-1GEA 41.2 USD
TCM2874-1G TCM2874-5G
4-(Methylthio)phenylacetic acid ≥98.0% (by GC, titration analysis)
4-(Methylthio)phenylacetic acid
Formula: C₉H₁₀O₂S
MW: 182.24
MDL Number: MFCD00192325
CAS Number: 16188-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41442a68515d60a38021079968f570a3cf830e5e&vid=debaa015c437a06ec8fa330f93534af27b7014ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR