4-Methoxycarbonyl-2-nitrophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-(Methoxycarbonyl)-2-nitrophenylboronic acid, 4-(Methoxycarbonyl)-2-nitrobenzeneboronic acid, 4-Methoxycarbonyl-2-nitrobenzeneboronic acid

M2854-1G M2854-5G
TCM2854-1GEA 44.29 USD
TCM2854-1G TCM2854-5G
4-Methoxycarbonyl-2-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Methoxycarbonyl-2-nitrophenylboronic acid
Formula: C₈H₈BNO₆
Melting Pt: 168 °C
MDL Number: MFCD01632202
CAS Number: 85107-55-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0d2b06c6a9eb9845a85268e6324e26af238ed31&vid=8af26cd965475e7df26b2f67ec62fcf7462ac7ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR