Methyl 2-Fluoro-2-methylpropionate ≥95.0% (by GC)

Supplier: TCI America
Danger

M2827-25G M2827-5G
TCM2827-25GEA 231.75 USD
TCM2827-25G TCM2827-5G
Methyl 2-Fluoro-2-methylpropionate ≥95.0% (by GC)
Methyl 2-Fluoro-2-methylpropionate
Formula: C₅H₉FO₂
Boiling Pt: 109 °C
MDL Number: MFCD03265444
CAS Number: 338-76-1
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0aa4d9172775c9c3ccf4255814c20eede50c8aa&vid=044eff8a6dc4ac47bad289300360edb7279ba676&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR