1-Methyl-3-phenyl-2-thiourea ≥97.0% (by HPLC, total nitrogen)
Supplier: TCI America
|
Danger
|
Synonyms:
1-Methyl-3-phenylthiourea
M2786-1G
M2786-5G
TCM2786-1GEA
87.55
USD
TCM2786-1G
TCM2786-5G
1-Methyl-3-phenyl-2-thiourea ≥97.0% (by HPLC, total nitrogen)
1-Methyl-3-phenyl-2-thiourea
Formula:
C₈H₁₀N₂S |
MDL Number:
MFCD00022130 CAS Number: 2724-69-8 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...