Isodecyl acrylate (mixture of branched chain isomers) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

I1104-100G I1104-25G
TCI1104-100GEA 123.6 USD
TCI1104-100G TCI1104-25G
Isodecyl acrylate (mixture of branched chain isomers) stabilized
Isodecyl acrylate (mixture of branched chain isomers)
Formula: C₁₃H₂₄O₂
Boiling Pt: 158 °C
Flash Pt: 116 °C
MDL Number: MFCD00080455
CAS Number: 1330-61-6
EINECS: 215-542-5
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caa9c96bad2b26208e4310ff8ca7cc9570c1e64c&vid=eb36f9c4fb45cb56b250ab549da53681ea76d2d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) many component mixture


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR