1,1'-(Hexane-1,6-diyl)bis[3-(2-nitrophenyl)urea] ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1-(2-Nitrophenyl)-3-[6-[(2-nitrophenyl)carbamoylamino]hexyl]urea

H1655-1G H1655-5G
TCH1655-1GEA 121.54 USD
TCH1655-1G TCH1655-5G
1,1'-(Hexane-1,6-diyl)bis[3-(2-nitrophenyl)urea] ≥98.0% (by HPLC, total nitrogen)
1,1'-(Hexane-1,6-diyl)bis[3-(2-nitrophenyl)urea]
Formula: C₂₀H₂₄N₆O₆
MW: 444.45 g/mol
CAS Number: 2419249-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=875aa2e29d07e3d978129d6a26f81b42d6483200&vid=7717ac2696b9d431396ebee23e24c203638252a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR