N-Hydroxynaphthalimide trifluoromethanesulfonate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1,3-Dioxo-1H-benzo[de]isoquinolin-2(3H)-yl trifluoromethanesulphonate, N-(Trifluoromethylsulphonyloxy)-1,8-naphthalimide, N-Hydroxynaphthalimide triflate

H1136-1G H1136-250MG
TCH1136-1GEA 166.86 USD
TCH1136-1G TCH1136-250MG
N-Hydroxynaphthalimide trifluoromethanesulfonate ≥98.0% (by HPLC)
N-Hydroxynaphthalimide trifluoromethanesulfonate
Formula: C₁₃H₆F₃NO₅S
MW: 345.25 g/mol
MDL Number: MFCD02683478
CAS Number: 85342-62-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 211.0 to 215.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR