4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: trans,trans-4'-Pentylbicyclohexyl-4-carboxylic acid 4-Fluorophenyl ester, 4-Fluorophenyl trans,trans-4'-Pentylbicyclohexyl-4-carboxylate, (Trans,trans)-4-fluorophenyl 4'-pentyl-[1,1'-bi(cyclohexane)]-4-carboxylate

F1120-1G F1120-5G
TCF1120-1GEA 96.82 USD
TCF1120-1G TCF1120-5G
4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate ≥98.0% (by GC)
4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate
Formula: C₂₄H₃₅FO₂
MW: 374.54 g/mol
MDL Number: MFCD11053423
CAS Number: 88878-50-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR