5-Chloro-2-fluorobenzotrifluoride ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Chloro-4-fluoro-3-(trifluoromethyl)benzene, 2-Fluoro-5-chlorobenzotrifluoride, 4-Chloro-1-fluoro-2-(trifluoromethyl)benzene, 5-Chloro-2-fluorobenzotrifluoride

C3317-1G C3317-5G
TCC3317-1GEA 43.26 USD
TCC3317-1G TCC3317-5G
5-Chloro-2-fluorobenzotrifluoride ≥98.0% (by GC)
5-Chloro-2-fluorobenzotrifluoride
Formula: C₇H₃ClF₄
MW: 198.55 g/mol
Flash Pt: 47 °C
MDL Number: MFCD00060655
CAS Number: 89634-74-2
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=574dff591d7d8c68a4d862742149b0e9e310bfce&vid=57ee872c057927cf45e89aee4fb902db435ebe6a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR