5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-2-triphenylmethyl-2H-tetrazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-2-trityl-2H-tetrazole

B5933-1G B5933-5G
TCB5933-1GEA 83.43 USD
TCB5933-1G TCB5933-5G
5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-2-triphenylmethyl-2H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-[4'-(Bromomethyl)-1,1'-biphenyl-2-yl]-2-triphenylmethyl-2H-tetrazole
Formula: C₃₃H₂₅BrN₄
Melting Pt: 137 °C
MDL Number: MFCD08274767
CAS Number: 133051-88-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bebeb87cc9aa96f4585eccc0baf028d96433ccbd&vid=926dd9bbb9e5e44b3b91f6ecf0050445f4d672ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR