9-Bromo-10-[3-(2-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)

Supplier: TCI America

B5772-1G B5772-200MG
TCB5772-1GEA 332.69 USD
TCB5772-1G TCB5772-200MG
9-Bromo-10-[3-(2-naphthyl)phenyl]anthracene ≥98.0% (by HPLC)
9-Bromo-10-[3-(2-naphthyl)phenyl]anthracene
Formula: C₃₀H₁₉Br
MDL Number: MFCD30470616
CAS Number: 944801-33-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11e652e371cbde9a82cc80b3ca1c63a8fd65c18d&vid=fe67542a10c525972744be161a1691e1e59e746b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR